Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Soap Disp SLVR 800 mL 1 600 mL 6 inD

Catalog No. 502758468
Encompass_Preferred

Soap Disp SLVR 800 mL 1 600 mL 6 inD

Catalog No. 50-275-8468 Supplier Grainger Supplier No. 15A649
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Soap Disp SLVR 800 mL 1 600 mL 6 inD

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.