Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Soap Power!
GREENER_CHOICESCI_ED

Catalog No. S42856 Shop All Aldon Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S42856 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CHOKING HAZARD - Small parts. Not for children under 3 yrs.
Catalog No. S42856 Supplier Innovating Science™ Supplier No. IS1006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHOKING HAZARD - Small parts. Not for children under 3 yrs.

Demonstrate the concept of surface tension to your classroom using soap as a power source!

  • Why are raindrops the shape that they are? How do detergents work? Simply put - surface tension
  • The Soap Power! Kit contains materials to demonstrate the concept of surface tension to your classroom using soap as a power source
  • Kit contains enough materials for 6 groups of 4
  • Developed for grades 2 through 5

Specifications

Certifications/Compliance Product SDSs
Color White
Grade Level Elementary
Grade 2 to 5
Age 10 to 13
For Use With (Application) Science Education
Includes 6 x 5 mL Dropper Bottle, Liquid Dish Soap, 2 Foam Trays, 6 Plastic Trays, 6 Pepper Packets
Quantity 1
Weight (English) 2 lb.
Dimensions (L x W x H) 10 x 7 x 5 in.
Class Size 6 Lab Groups
Product Type General Chemistry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.