Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Soap Solution, Hardness Demonstration, 500 mL

Catalog No. 501994308 Shop All Hach Company Products
Encompass

Soap solution for hardness demonstration.- 25 mL sample. 500 mL bottle

Catalog No. 50-199-4308 Supplier Hach Company Supplier No. 46249
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Soap solution for hardness demonstration.- 25 mL sample. 500 mL bottle

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.