Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Soap Solution, Hardness Demonstration, 1 L

Catalog No. 501994269 Shop All Hach Company Products
Encompass

Soap solution for Hardness Demonstration. 50ml sample. 1000 mL bottle

Catalog No. 50-199-4269 Supplier Hach Company Supplier No. 39553
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Soap solution for Hardness Demonstration. 50ml sample. 1000 mL bottle

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.