Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soap Solution for Biodiesel, (Acetone/Water/Bromophenol Blue), Reagents

Catalog No. 01333911
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-911 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-911 Supplier Reagents Holdings Llc Supplier No. CS02700020E7
Only null left
Add to Cart
Add to Cart

Soap Solution for Biodiesel, (Acetone/Water/Bromophenol Blue), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Soap Solution for Biodiesel, (Acetone/Water/Bromophenol Blue)
Color Green
CAS 67-64-1, 7732-18-5, 64-17-5
Physical Form Liquid
Quantity 20 L
Packaging HDPE Winpak
Specific Gravity 0.7906
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.