Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soap Solution for Oils, (Acetone/Bromophenol Blue), Reagents

Catalog No. 01333908
Change view
Click to view available options
Packaging:
Cubitainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
01-333-908 Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-908 Supplier Reagents Holdings Llc Supplier No. CS02720020F
Add to Cart
Add to Cart

Soap Solution for Oils, (Acetone/Bromophenol Blue), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Soap Solution for Oils, (Acetone/Bromophenol Blue)
Color Blue
CAS 67-64-1, 7732-18-5
Physical Form Liquid
Boiling Point 56°C
Quantity 20 L
Packaging Cubitainer
Specific Gravity 0.7935
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.