Learn More
Enterprise Technology Solutions Soap2o Hand Soap Kit,w/Dispenser/HydroDrop,Foaming,Coconut Acai,AR
SDP
Catalog No. 5000828749
Encompass_Preferred
HydroDrops Soap Kit includes one dispenser and two HydroDrops. Just one HydroDrop creates half a liter of luxurious, foaming hand soap that fits perfectly in the included pump bottle. Dissolvable drops are made with naturally derived, vegan-friendly ingredients. HydroDrops eliminate the shipping of water and plastic waste. Each drop supports Soap2o's mission to provide 16 liters of fresh drinking water to communities in developing countries via the Made Blue Foundation.
Catalog No. 50-008-28749 Supplier Enterprise Technology Solutions Supplier No. SOAS2OKIT
HydroDrops Soap Kit includes one dispenser and two HydroDrops. Just one HydroDrop creates half a liter of luxurious, foaming hand soap that fits perfectly in the included pump bottle. Dissolvable drops are made with naturally derived, vegan-friendly ingredients. HydroDrops eliminate the shipping of water and plastic waste. Each drop supports Soap2o's mission to provide 16 liters of fresh drinking water to communities in developing countries via the Made Blue Foundation.