Learn More
Alkali Scientific SOB Broth, 1 Kg
SDP
Catalog No. 5000660996
Encompass_Preferred
SOB Broth (Standard Oxytetracycline Broth) is a microbiological growth medium used to support the growth of a variety of bacterial species. The 1 kg package is ideal for large-scale microbial growth studies and testing in clinical, pharmaceutical, and environmental laboratories. This broth is particularly effective in the recovery and cultivation of organisms from water and environmental samples. The medium contains a broad-spectrum antibiotic (oxytetracycline) that suppresses unwanted bacterial growth, allowing researchers to focus on the target organisms. SOB Broth is commonly used in research, diagnostics, and quality control testing to isolate and enumerate specific bacterial populations from contaminated samples.
Catalog No. 50-006-60996 Supplier Alkali Scientific Supplier No. SOB003CP
SOB Broth (Standard Oxytetracycline Broth) is a microbiological growth medium used to support the growth of a variety of bacterial species. The 1 kg package is ideal for large-scale microbial growth studies and testing in clinical, pharmaceutical, and environmental laboratories. This broth is particularly effective in the recovery and cultivation of organisms from water and environmental samples. The medium contains a broad-spectrum antibiotic (oxytetracycline) that suppresses unwanted bacterial growth, allowing researchers to focus on the target organisms. SOB Broth is commonly used in research, diagnostics, and quality control testing to isolate and enumerate specific bacterial populations from contaminated samples.