Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp SOB Broth, 1 Kilogram, Nutritionally Rich Growth Medium
SDP

Catalog No. 501977597
Encompass_Preferred

SOB Broth is recommended for use in cultivation of recombinant strains of E. Coli. It is a nutritionally rich growth medium for use in the preparation and transformation of competent cells.

Add 28 grams of Broth in 1 liter of distilled water. Heat gently to dissolve the medium completely. Dispense in tubes and sterilize by autoclaving for 15 minutes.

Ingredients:

Casein Enzymic Hydrolysate: 20 g/l
Magnesium Sulfate: 2.4 g/l
Yeast Extract: 5 g/l
Potassium Chloride: 0.186 g/l
Sodium Chloride: 0.5 g/l
Final pH(25°:C): 7.0+0.2

Catalog No. 50-197-7597 Supplier Research Products International Corp Supplier No. S250001000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SOB Broth is recommended for use in cultivation of recombinant strains of E. Coli. It is a nutritionally rich growth medium for use in the preparation and transformation of competent cells.

Add 28 grams of Broth in 1 liter of distilled water. Heat gently to dissolve the medium completely. Dispense in tubes and sterilize by autoclaving for 15 minutes.

Ingredients:

Casein Enzymic Hydrolysate: 20 g/l
Magnesium Sulfate: 2.4 g/l
Yeast Extract: 5 g/l
Potassium Chloride: 0.186 g/l
Sodium Chloride: 0.5 g/l
Final pH(25°:C): 7.0+0.2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.