Learn More
Teknova SOB Broth Medium. 227 grams.
SDP
Catalog No. 50843062
Encompass
SOB (Super Optimal Broth) Medium is a nutrient rich medium for cultivation ofE. coli, often used to give high transformation efficiency. SOC Medium is used for providing nutrients and supporting the recovery of Escherichia coli competent cells following transformation.
Catalog No. 50-843-062 Supplier Teknova Supplier No. S0221
SOB (Super Optimal Broth) Medium is a nutrient rich medium for cultivation ofE. coli, often used to give high transformation efficiency. SOC Medium is used for providing nutrients and supporting the recovery of Escherichia coli competent cells following transformation.