Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp SOB Broth, 36 x 500ml Capsules, 500 Grams
SDP

Catalog No. 50489040
Encompass_Preferred

SOB Broth is recommended for use in cultivation of recombinant strains of E. Coli. It is a nutritionally rich growth medium for use in the preparation and transformation of competent cells.

Add two capsules to one liter of distilled water. Heat and stir gently to completely dissolve medium. Sterilize by autoclaving at 121°C for 15 minutes.

Final Solution contains:

Casein Enzymic Hydrolysate: 20 g/l
Yeast Extract: 5 g/l
Sodium Chloride: 0.5 g/l
Magnesium Sulfate: 2.4 g/l
Potassium Chloride: 0.186 g/l

Catalog No. 50-489-040 Supplier Research Products International Corp Supplier No. S25040500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SOB Broth is recommended for use in cultivation of recombinant strains of E. Coli. It is a nutritionally rich growth medium for use in the preparation and transformation of competent cells.

Add two capsules to one liter of distilled water. Heat and stir gently to completely dissolve medium. Sterilize by autoclaving at 121°C for 15 minutes.

Final Solution contains:

Casein Enzymic Hydrolysate: 20 g/l
Yeast Extract: 5 g/l
Sodium Chloride: 0.5 g/l
Magnesium Sulfate: 2.4 g/l
Potassium Chloride: 0.186 g/l

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.