Learn More
Alkali Scientific SOB Broth, 72 x 500mL Capsules, 1 Kg
SDP
Catalog No. 5000661147
Encompass_Preferred
SOB Broth (72 x 500mL Capsules, 1 Kg) is a high-nutrient microbial culture medium formulated to promote rapid bacterial growth. The SOB formula is designed for applications that require optimal conditions for bacterial propagation, supporting a wide range of bacterial species. This product includes 72 buffered capsules, each containing a precise quantity of nutrients necessary to prepare 500mL of medium. SOB Broth is widely used in microbiology labs for culturing a variety of bacterial strains, performing antimicrobial testing, and supporting routine microbiological research. The convenient capsule form ensures accurate, easy preparation, and eliminates the need for manual measurement.
Catalog No. 50-006-61147 Supplier Alkali Scientific Supplier No. SOB001CP
SOB Broth (72 x 500mL Capsules, 1 Kg) is a high-nutrient microbial culture medium formulated to promote rapid bacterial growth. The SOB formula is designed for applications that require optimal conditions for bacterial propagation, supporting a wide range of bacterial species. This product includes 72 buffered capsules, each containing a precise quantity of nutrients necessary to prepare 500mL of medium. SOB Broth is widely used in microbiology labs for culturing a variety of bacterial strains, performing antimicrobial testing, and supporting routine microbiological research. The convenient capsule form ensures accurate, easy preparation, and eliminates the need for manual measurement.