Learn More
Biotang Inc General Molecular Biology Grade Water, WT-020, 500 ml. General Molecular Biology Grade - Sterile Certified Nuclease Free Endotoxin Free<0.01EU/ml Method Distilled and steam sterilized at121 0C/15PSI for 60 min
Catalog No. 50155892 Shop All BIOTANG Inc Products
Encompass_Preferred
General Molecular Biology Grade Water, WT-020, 500 ml. General Molecular Biology Grade - Sterile Certified Nuclease Free Endotoxin Free<0.01EU/ml Method Distilled and steam sterilized at121 0C/15PSI for 60 min.
Catalog No. 50-155-892 Supplier Biotang Inc Supplier No. BTBB570
General Molecular Biology Grade Water, WT-020, 500 ml. General Molecular Biology Grade - Sterile Certified Nuclease Free Endotoxin Free<0.01EU/ml Method Distilled and steam sterilized at121 0C/15PSI for 60 min.