Learn More
Medchemexpress LLC HY-14823 10mg Medchemexpress, Sobetirome CAS:211110-63-3 Purity:>98%
SDP
Catalog No. 502029070
Encompass_Preferred
Medchemexpress, HY-14823 10mg Sobetirome CAS:211110-63-3 Sobetirome (GC-1) is a thyroid hormone receptor β (TRβ)-specific agonist which bind selectively to TRβ-1 with an EC50 of 0.16 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9070 Supplier Medchemexpress LLC Supplier No. HY1482310MG
Medchemexpress, HY-14823 10mg Sobetirome CAS:211110-63-3 Sobetirome (GC-1) is a thyroid hormone receptor β (TRβ)-specific agonist which bind selectively to TRβ-1 with an EC50 of 0.16 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.