Learn More
Medchemexpress LLC HY-14672 5mg Medchemexpress, Soblidotin CAS:149606-27-9 Purity:>98%
SDP
Catalog No. 502081629
Encompass_Preferred
Medchemexpress, HY-14672 5mg Soblidotin CAS:149606-27-9 Soblidotin (Auristatin PE) is a novel synthetic Dolastatin 10 derivative and inhibitor of tubulin polymerization. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1629 Supplier Medchemexpress LLC Supplier No. HY146725MG
Medchemexpress, HY-14672 5mg Soblidotin CAS:149606-27-9 Soblidotin (Auristatin PE) is a novel synthetic Dolastatin 10 derivative and inhibitor of tubulin polymerization. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.