Learn More
Kd Medical Inc SOC MEDIA 10 X 10ML
SDP
Catalog No. 501018939
Encompass_Preferred
SOC is a nutritionally rich medium used to grow recombinant E.coli.
Catalog No. 50-101-8939 Supplier Kd Medical Inc Supplier No. BLA5140
SOC is a nutritionally rich medium used to grow recombinant E.coli.