Learn More
TRIBIOSCIENCE INC SOC Medium, 100 ML
SDP
Catalog No. NC3834045
Encompass
Tribioscience SOC Medium is used in the final step of bacterial cell transformation to obtain maximal transformation efficiency of E. coli. This medium is supplied as 100mL bottles of liquid medium.
Catalog No. NC3834045 Supplier TRIBIOSCIENCE INC Supplier No. TBS8058
Tribioscience SOC Medium is used in the final step of bacterial cell transformation to obtain maximal transformation efficiency of E. coli. This medium is supplied as 100mL bottles of liquid medium.