Learn More
Teknova SOC Medium, 1000mL, Sterile.
SDP
Catalog No. NC9765471
Encompass
2.00% Tryptone, 0.50% Yeast Extract, 10mM Sodium Chloride, 2.5mM Potassium Chloride, 10mM Magnesium Chloride, 10mM Magnesium Sulfate, 20mM Cerelose Anhydrous Dextrose (Glucose)
Catalog No. NC9765471 Supplier Teknova Supplier No. S1660
2.00% Tryptone, 0.50% Yeast Extract, 10mM Sodium Chloride, 2.5mM Potassium Chloride, 10mM Magnesium Chloride, 10mM Magnesium Sulfate, 20mM Cerelose Anhydrous Dextrose (Glucose)