Learn More
Teknova SOC Medium, 100mL. Sterile.
SDP
Catalog No. 501908117
Encompass
2.0% Tryptone, 0.5% Yeast Extract, 10mM Sodium Chloride, 2.5mM Potassium Chloride, 10mM Magnesium Chloride, 10mM Magnesium Sulfate, 20mM Cerelose Anhydrous Dextrose (Glucose)
Catalog No. 50-190-8117 Supplier Teknova Supplier No. S1630
2.0% Tryptone, 0.5% Yeast Extract, 10mM Sodium Chloride, 2.5mM Potassium Chloride, 10mM Magnesium Chloride, 10mM Magnesium Sulfate, 20mM Cerelose Anhydrous Dextrose (Glucose)