Learn More
Teknova SOC Medium. 20mL, Sterile.
SDP
Catalog No. 502388070
Encompass
2.0% Tryptone, 0.5% Yeast Extract,10mM Sodium Chloride, 2.5mM Potassium Chloride,10mM Magnesium Chloride, 10mM Magnesium Sulfate,20mM Cerelose Anhydrous Dextrose (Glucose)
Catalog No. 50-238-8070 Supplier Teknova Supplier No. S1642
2.0% Tryptone, 0.5% Yeast Extract,10mM Sodium Chloride, 2.5mM Potassium Chloride,10mM Magnesium Chloride, 10mM Magnesium Sulfate,20mM Cerelose Anhydrous Dextrose (Glucose)