Learn More
Teknova SOC Medium. 7mL, Sterile. 10-pack 10 pack.
SDP
Catalog No. 502387560
Encompass
2.0% Tryptone, 0.5% Yeast Extract, 10mM Sodium Chloride, 2.5mM Potassium Chloride, 10mM Magnesium Chloride, 10mM Magnesium Sulfate, 20mM Cerelose Anhydrous Dextrose (Glucose)
Catalog No. 50-238-7560 Supplier Teknova Supplier No. S165710PK
2.0% Tryptone, 0.5% Yeast Extract, 10mM Sodium Chloride, 2.5mM Potassium Chloride, 10mM Magnesium Chloride, 10mM Magnesium Sulfate, 20mM Cerelose Anhydrous Dextrose (Glucose)