Learn More
Sigma Aldrich Fine Chemicals Biosciences SOC Medium for use in tran
Catalog No. 502900667
Encompass
SOC is a microbial growth medium used for the transformation of competent cells (E. coli). This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and glucose in a low-salt formulation.
Catalog No. 50-290-0667 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S1797100ML
SOC is a microbial growth medium used for the transformation of competent cells (E. coli). This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and glucose in a low-salt formulation.