Learn More
Sigma Aldrich Fine Chemicals Biosciences SOC Medium for use in tran
Supplier: Sigma Aldrich Fine Chemicals Biosciences S179710X5ML
SOC is a microbial growth medium used for the transformation of competent cells (E. coli). This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and glucose in a low-salt formulation.