Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOC medium

Catalog No. AAJ60255AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60255AE 100 mL
AAJ60255AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60255AE Supplier Thermo Scientific Chemicals Supplier No. J60255AE
Add to Cart

For cultivating recombinant strains of Escherichia coli. SOC Medium is used in the final step of bacterial cell transformation to obtain the maximal transformation efficiency of E. coli.

For cultivating recombinant strains of Escherichia coliSOC Medium is used in the final step of bacterial cell transformation to obtain the maximal transformation efficiency of E. coli.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00284637

Specifications

Packaging Plastic bottle
Quantity 100 mL
Solubility Information Miscible with water.
Physical Form Liquid
Chemical Name or Material SOC medium
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.