Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SOCE inhibitor 1 | 2169316-15-6 | 99.7% | 513.47 | 100 MG
SDP

Catalog No. 5000439662
Encompass_Preferred

SOCE inhibitor 1 (Compound 39) is a store-operated calcium entry (SOCE) inhibitor with an IC50 of 4.4 μM. It demonstrates good stability in plasma. This product is for research use only and is not sold to patients.

  • Target: store-operated calcium entry (SOCE)
  • IC50: 4.4 μM for SOCE
  • Stability: good stability in plasma
  • Appearance: solid, white to pink
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-39662 Supplier Medchemexpress LLC Supplier No. HY112913100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

SOCE inhibitor 1 (Compound 39) is a store-operated calcium entry (SOCE) inhibitor with an IC50 of 4.4 μM. It demonstrates good stability in plasma. This product is for research use only and is not sold to patients.

  • Target: store-operated calcium entry (SOCE)
  • IC50: 4.4 μM for SOCE
  • Stability: good stability in plasma
  • Appearance: solid, white to pink
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.