Learn More
CEM Corporation SOCKET 19MM 6PT
SDP
Catalog No. NC0941206
Encompass
Socket for Torque Wrench, 19 mm, 6 point. Use with Torque Wrench, Part 404070 or 432201
Catalog No. NC0941206 Supplier CEM Corporation Supplier No. 432195
Socket for Torque Wrench, 19 mm, 6 point. Use with Torque Wrench, Part 404070 or 432201