Learn More
MSC Proto Hex Bit Socket, 3/8 in, 5 mm, 1 ea
Catalog No. 501920484
Encompass
Proto hex bit sockets are engineered for high-performance applications, constructed from a heat-treated alloy for wear-resistance which helps minimize distortion for a long service life. The hex opening is designed to securely hold the bit, allowing the bit holder to absorb the torque load effectively.
  • Constructed from chrome alloy steel
  • Features a polished chrome finish with a black oxide stem
  • Designed to minimize distortion for extended tool life
  • Hex opening ensures secure bit retention and optimal torque transfer
Catalog No. 50-192-0484 Supplier MSC Supplier No. 39937388
Proto hex bit sockets are engineered for high-performance applications, constructed from a heat-treated alloy for wear-resistance which helps minimize distortion for a long service life. The hex opening is designed to securely hold the bit, allowing the bit holder to absorb the torque load effectively.
  • Constructed from chrome alloy steel
  • Features a polished chrome finish with a black oxide stem
  • Designed to minimize distortion for extended tool life
  • Hex opening ensures secure bit retention and optimal torque transfer