Learn More
MSC Proto Hex Bit Socket, 1/2 in, 9/16 in, 1 ea
Catalog No. 501920471
Encompass
This hand hex bit socket is designed for precision fastening and unfastening tasks. Crafted from durable chrome alloy steel, it features a 1/2 in drive and a 9/16 in hex size, making it suitable for various mechanical applications.
  • Socket type: hex bit
  • Drive size: 1/2 in
  • Hex size: 9/16 in
  • Bit length: 1-7/8 in
  • Overall length: 6-29/32 in
  • Material: chrome alloy steel
  • Finish: black oxide (stem); polished chrome
  • Not insulated
  • Not tether capable
  • Non-sparking
Catalog No. 50-192-0471 Supplier MSC Supplier No. 39937669
This hand hex bit socket is designed for precision fastening and unfastening tasks. Crafted from durable chrome alloy steel, it features a 1/2 in drive and a 9/16 in hex size, making it suitable for various mechanical applications.
  • Socket type: hex bit
  • Drive size: 1/2 in
  • Hex size: 9/16 in
  • Bit length: 1-7/8 in
  • Overall length: 6-29/32 in
  • Material: chrome alloy steel
  • Finish: black oxide (stem); polished chrome
  • Not insulated
  • Not tether capable
  • Non-sparking