Learn More
Ampco Safety Tools Standard Socket, 1/2 in. Drive (Metric)
SDP
Catalog No. 19174420 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (Metric):
6 mm
7 mm
8 mm
9 mm
10 mm
11 mm
12 mm
13 mm
15 mm
18 mm
19 mm
20 mm
21 mm
22 mm
23 mm
24 mm
25 mm
26 mm
27 mm
28 mm
29 mm
30 mm
31 mm
32 mm
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
19-174-420 21 mm
19-174-398 10 mm
19-174-403 11 mm
19-174-404 12 mm
19-174-408 13 mm
19-174-412 15 mm
19-174-414 18 mm
19-174-417 19 mm
19-174-418 20 mm
19-174-421 22 mm
19-174-422 23 mm
19-174-423 24 mm
19-174-425 25 mm
19-174-426 26 mm
19-174-427 27 mm
19-174-428 28 mm
19-174-429 29 mm
19-174-432 30 mm
19-174-433 31 mm
19-174-434 32 mm
19-174-436 6 mm
19-174-439 7 mm
19-174-440 8 mm
19-174-441 9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-174-420 Supplier Ampco Safety Tools Supplier No. SS1/2D21MM
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Standard Socket performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type Standard Socket Wrench
Drive Size 1/2 in.
Length (Metric) 21 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.