Learn More
MSC Proto Hex Bit Socket, 3/8 in, 10 mm, 1 ea
Catalog No. 501920488
Encompass
This hex bit socket is designed for hand use, featuring a 3/8 in drive and a 10.00 mm hex size. Constructed from chrome alloy steel, it offers durability and reliability for various applications.
  • Hex bit socket type
  • Drive size of 3/8 in
  • Hex size of 10.00 mm
  • Chrome alloy steel material
  • Black oxide (stem) and polished chrome finish
  • Bit length of 1-5/8 in
  • Overall length of 6-29/32 in
  • Not insulated
  • Not tether capable
  • Manufactured in the United States
Catalog No. 50-192-0488 Supplier MSC Supplier No. 39937347
This hex bit socket is designed for hand use, featuring a 3/8 in drive and a 10.00 mm hex size. Constructed from chrome alloy steel, it offers durability and reliability for various applications.
  • Hex bit socket type
  • Drive size of 3/8 in
  • Hex size of 10.00 mm
  • Chrome alloy steel material
  • Black oxide (stem) and polished chrome finish
  • Bit length of 1-5/8 in
  • Overall length of 6-29/32 in
  • Not insulated
  • Not tether capable
  • Manufactured in the United States