Learn More
MSC Proto Hex Bit Socket, 3/8 in, 5/16 in, 1 ea
Catalog No. 501920481
Encompass
This hand hex bit socket is designed for use with 3/8 in drive tools, featuring a 5/16 in hex size. Constructed from chrome alloy steel, it is suitable for various fastening applications.
  • Drive size: 3/8 in
  • Hex size: 5/16 in
  • Bit length: 1-5/8 in
  • Overall length: 6-29/32 in
  • Material: chrome alloy steel
  • Finish: black oxide (stem); polished chrome
  • Not insulated
  • Not tether capable
Catalog No. 50-192-0481 Supplier MSC Supplier No. 39937479
This hand hex bit socket is designed for use with 3/8 in drive tools, featuring a 5/16 in hex size. Constructed from chrome alloy steel, it is suitable for various fastening applications.
  • Drive size: 3/8 in
  • Hex size: 5/16 in
  • Bit length: 1-5/8 in
  • Overall length: 6-29/32 in
  • Material: chrome alloy steel
  • Finish: black oxide (stem); polished chrome
  • Not insulated
  • Not tether capable