Learn More
DWK LIFE SCIENCES™ Socorex™ Micropipette Rack for Qualitix™ Tipfill Refill System
Shop All DWK Life Sciences Products

Description
- Accepts Qualitix™ Tipfill Refill System refill packs.
- Hinged lid.
- Smooth, rounded edges.
- Ergonomic snap latch that won't catch gloves.
- Transparent lid.
- Reusable.
Specifications
Specifications
| Description | Empty rack for 10/200 μL |
| Autoclavable | Autoclavable |
| Color | Natural |
| Holds | Micropipette Tips (200 μL ) |
| Material | Polypropylene |
| Positions | 96 |
| Shape | Rectangular |
| Stackable | Yes |
| Sterility | Non-sterile |
| Packaging | 10 Pk. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.