Learn More
Aviva Systems Biology Corp SOCS1 Antibody - middle region (ARP42148_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42148_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOCS1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SOCS1. Alternate names: CIS1, CISH1, JAB, SOCS-1, SSI-1, SSI1, TIP3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-980