Learn More
Aviva Systems Biology Corp SOCS7 Antibody - N-terminal region (ARP53700_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53700_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOCS7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SOCS7. Alternate names: NAP4, NCKAP4ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61098