Learn More
McKesson General Medical freeflex® Sodium Chloride 0.9% IV Solution 250 mL, 30/cs
Catalog No. NC2953687
Encompass
0.9% Sodium Chloride InjectionThe unique, blue-and-white ports are designed with finger guards for easy handlingBoth ports contain easy break-off caps that provide visual tamper evidence and ensure sterility prior to useThe overwrap is made of polypropylene filmThe freeflex® bag is a multilayer polyolefin filmProminent labeling allows differentiation between products and fill sizesPlease refer to the package insert for the applicable product for a list of precautions and warnings
Catalog No. NC2953687 Supplier McKesson General Medical Supplier No. 1104661
0.9% Sodium Chloride InjectionThe unique, blue-and-white ports are designed with finger guards for easy handlingBoth ports contain easy break-off caps that provide visual tamper evidence and ensure sterility prior to useThe overwrap is made of polypropylene filmThe freeflex® bag is a multilayer polyolefin filmProminent labeling allows differentiation between products and fill sizesPlease refer to the package insert for the applicable product for a list of precautions and warnings