Learn More
Med Vet International Sodium Citrate 4% Anticoagulant 250mL, J0521
Catalog No. 502828936
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Sodium Citrate. A very familiar product to veterinarians. It has no RBC preservative, and thus should be used for blood therapy that will be done with a short 24 to 48-hour period of time. If only the plasma component is to be used it will also work well for that purpose. Sodium Citrate 4% Anticoagulant. 250ml. Great for use in equine plasma therapy or small animal short-term whole blood therapy. Dilute 1ml to 79ml of blood
Catalog No. 50-282-8936 Supplier Med Vet International Supplier No. RXSODCIT
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Sodium Citrate. A very familiar product to veterinarians. It has no RBC preservative, and thus should be used for blood therapy that will be done with a short 24 to 48-hour period of time. If only the plasma component is to be used it will also work well for that purpose. Sodium Citrate 4% Anticoagulant. 250ml. Great for use in equine plasma therapy or small animal short-term whole blood therapy. Dilute 1ml to 79ml of blood