Learn More
Bioworld Sodium Deoxycholate, 10% , 500 mL
Catalog No. NC0839985
Encompass_Preferred
Deoxycholic Acid, Sodium Salt (Sodium Deoxycholate) An anionic detergent extracted from bovine bile. High Purity Grade sodium deoxycholic acid dissolved at 10% stock solution.Limited shelf life.An ionic detergent for the solubilization of membrane-bound proteinsKeep away from light.RTECS Number: FZ2250000
Catalog No. NC0839985 Supplier Bioworld Supplier No. 404300184
Deoxycholic Acid, Sodium Salt (Sodium Deoxycholate) An anionic detergent extracted from bovine bile. High Purity Grade sodium deoxycholic acid dissolved at 10% stock solution.Limited shelf life.An ionic detergent for the solubilization of membrane-bound proteinsKeep away from light.RTECS Number: FZ2250000