Learn More
IBI SCIENTIFIC 3P ULTRAPURE SDS POWDER (SODIUM DODECYL SULFATE) 1 KG
Catalog No. NC2829003
Encompass_Preferred
CAS# 151-21-3Ionic detergent used to denature proteinsAvailable in multiple sizes25 KG size must ship via truck on a palletBulk orders availableSodium Dodecyl Sulfate (SDS) is an ionic SDS detergent used to denature proteins in hybridization, nucleic acid purification, and electrophoresis buffer systems. SDS is also used to dissociate nucleic acid protein complexes in DNA extraction protocols, to disrupt cell membranes, and to prepare prehybridization and/or hybridization solutions.To make a 10% SDS solution, dissolve 100 g of SDS in 900 ml of ddi water. Heat to 68⁰ C to assist dissolution. Adjust the pH to 7.2 by adding a few drops of HCl. Adjust the total volume to 1 L with ddi water, then dispense into aliquots."
Catalog No. NC2829003 Supplier IBI SCIENTIFIC 3P Supplier No. IB07063
CAS# 151-21-3Ionic detergent used to denature proteinsAvailable in multiple sizes25 KG size must ship via truck on a palletBulk orders availableSodium Dodecyl Sulfate (SDS) is an ionic SDS detergent used to denature proteins in hybridization, nucleic acid purification, and electrophoresis buffer systems. SDS is also used to dissociate nucleic acid protein complexes in DNA extraction protocols, to disrupt cell membranes, and to prepare prehybridization and/or hybridization solutions.To make a 10% SDS solution, dissolve 100 g of SDS in 900 ml of ddi water. Heat to 68⁰ C to assist dissolution. Adjust the pH to 7.2 by adding a few drops of HCl. Adjust the total volume to 1 L with ddi water, then dispense into aliquots."