Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide - Chloride, 50%/1% (w/v), Ricca Chemical
SDP

Catalog No. R74781001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R74781001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R74781001A Supplier Ricca Chemical Company Supplier No. R74781001A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2 , 7647-14-5

Specifications

Physical Form Liquid
Quantity 1 L
Assay Percent Range 50/1% (w/v)
Grade Laboratory
Packaging Natural Poly Bottle
Chemical Name or Material Sod Hydrox-chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.