Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Potassium Tartrate, 0.25% (w/v), Ricca Chemical
SDP

Catalog No. R75350004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R75350004A 4 L
R75350001A 1 L
R753500020F 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R75350004A Supplier Ricca Chemical Company Supplier No. R75350004A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 6381-59-5
Molecular Weight (g/mol) 282.1 g/mol

Specifications

Physical Form Liquid
Quantity 4 L
Assay Percent Range 0.25% (w/v)
Percent Purity 0.25% W/V
Grade Laboratory
Packaging Natural Poly Bottle
Chemical Name or Material Sod Pot Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.