Learn More
Cayman Chemical SOD Stndard 1ea
SDP
Catalog No. 502896557
Encompass_Preferred
An antiparasitic agent reduces the proliferation of T. brucei (IC50 63.1 nM) but not T. cruzi L. infantum P. falciparum or human MRC-5 fibroblasts (IC50s >6000 nM for all) reduces peripheral blood parasite levels below detection limits and increases survival in a mouse model of T. brucei infection at 50 mg/kg twice per day
Catalog No. 50-289-6557 Supplier Cayman Chemical Supplier No. 7060051 EA
An antiparasitic agent reduces the proliferation of T. brucei (IC50 63.1 nM) but not T. cruzi L. infantum P. falciparum or human MRC-5 fibroblasts (IC50s >6000 nM for all) reduces peripheral blood parasite levels below detection limits and increases survival in a mouse model of T. brucei infection at 50 mg/kg twice per day