Learn More
Aviva Systems Biology Corp SOD1 Antibody - N-terminal region (ARP45752_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45752_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SOD1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SOD1. Alternate names: ALS, ALS1, IPOA, SOD, homodimer, hSod1ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-59128