Learn More
Montana Molecular SOD1 G85R BacMam - 5mL
SDP
Supplier:  Montana Molecular N0510N005
Encompass_Preferred
SOD1 with the G85R mutation packaged in BacMam for expression in mammalian cells. BacMam provides even, near 100% transduction efficiency in HEK293 cells as well as expression in many other cell lines, iPSCs, and primary cultures. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity. Wild-type or a version with a D90A mutation are also available. Co-express with Montana Molecular sensors for ER stress, cAMP, calcium, and more. This kit includes SOD1 G85R BacMam and 500mM sodium butyrate in H2O.
Catalog No. NC2600458