Learn More
Medchemexpress LLC SOD1 Human N-His 10ug
SDP
Catalog No. 5000196926
Encompass_Preferred
Recombinant human SOD1 protein with an N-terminal 6xHis tag, expressed in E. coli and supplied lyophilized. The protein is approximately 20.0 kDa and >95% pure by reducing SDS-PAGE, formulated in PBS (pH 7.4) with trehalose, mannitol, and Tween 80. Intended for biochemical assays, structural studies, and assay controls; store frozen and avoid reconstituting below recommended concentrations.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • N-terminal 6xHis tag for affinity purification or detection.
  • Lyophilized formulation for long-term stability.
  • Formulated in PBS with trehalose, mannitol, and Tween 80.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O.
  • Stable when stored at -20°C or -80°C; aliquot to avoid freeze-thaw.
  • Approximately 20.0 kDa molecular weight.
Catalog No. 50-001-96926 Supplier Medchemexpress LLC Supplier No. HYP71048A10UG
Recombinant human SOD1 protein with an N-terminal 6xHis tag, expressed in E. coli and supplied lyophilized. The protein is approximately 20.0 kDa and >95% pure by reducing SDS-PAGE, formulated in PBS (pH 7.4) with trehalose, mannitol, and Tween 80. Intended for biochemical assays, structural studies, and assay controls; store frozen and avoid reconstituting below recommended concentrations.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • N-terminal 6xHis tag for affinity purification or detection.
  • Lyophilized formulation for long-term stability.
  • Formulated in PBS with trehalose, mannitol, and Tween 80.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O.
  • Stable when stored at -20°C or -80°C; aliquot to avoid freeze-thaw.
  • Approximately 20.0 kDa molecular weight.