Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SOD1 Human N-His 1mg
SDP

Catalog No. 5000196851
Encompass_Preferred

Recombinant human Cu/Zn superoxide dismutase (SOD1) supplied as a lyophilized, active enzyme with an N-terminal 6xHis tag. The protein is expressed in E. coli, provided as a sterile powder, and formulated for stability in biochemical and enzymatic assays.

  • Lyophilized sterile powder formulation for long-term storage.
  • Formulated with trehalose, mannitol, and Tween 80 for enhanced stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Specific activity approximately 148.432 units/mg in a xanthine oxidase coupled assay.
  • Endotoxin content below 1 EU/μg, suitable for biochemical assays.
  • Stable frozen at -20°C; reconstituted material stable at 4°C for one week or longer if aliquoted and frozen.

Catalog No. 50-001-96851 Supplier Medchemexpress LLC Supplier No. HYP71048A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Cu/Zn superoxide dismutase (SOD1) supplied as a lyophilized, active enzyme with an N-terminal 6xHis tag. The protein is expressed in E. coli, provided as a sterile powder, and formulated for stability in biochemical and enzymatic assays.

  • Lyophilized sterile powder formulation for long-term storage.
  • Formulated with trehalose, mannitol, and Tween 80 for enhanced stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Specific activity approximately 148.432 units/mg in a xanthine oxidase coupled assay.
  • Endotoxin content below 1 EU/μg, suitable for biochemical assays.
  • Stable frozen at -20°C; reconstituted material stable at 4°C for one week or longer if aliquoted and frozen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.