Learn More
Medchemexpress LLC SOD1 Human N-His 500ug
SDP
Catalog No. 5000196868
Encompass_Preferred
Recombinant human superoxide dismutase 1 (SOD1) with an N-terminal 6xHis tag, supplied as a lyophilized powder produced in E. coli. The material is ≥97% pure (≈20 kDa), has low endotoxin (<1 EU/μg), and a measured specific activity of 148.432 U/mg. Lyophilized from PBS with trehalose and mannitol; store frozen and reconstitute according to guidance (do not reconstitute below 100 μg/mL).
  • High purity (~97%) suitable for biochemical and enzymatic assays.
  • N-terminal 6xHis tag for facile purification or detection.
  • Low endotoxin (<1 EU/μg) for sensitive biological applications.
  • Verified enzymatic activity (148.432 U/mg) for functional studies.
  • Lyophilized formulation with stabilizers for improved shelf life.
  • Expressed in E. coli to provide consistent recombinant supply.
  • Reconstitution guidance: avoid concentrations below 100 μg/mL in ddH2O.
Catalog No. 50-001-96868 Supplier Medchemexpress LLC Supplier No. HYP71048A500UG
Recombinant human superoxide dismutase 1 (SOD1) with an N-terminal 6xHis tag, supplied as a lyophilized powder produced in E. coli. The material is ≥97% pure (≈20 kDa), has low endotoxin (<1 EU/μg), and a measured specific activity of 148.432 U/mg. Lyophilized from PBS with trehalose and mannitol; store frozen and reconstitute according to guidance (do not reconstitute below 100 μg/mL).
  • High purity (~97%) suitable for biochemical and enzymatic assays.
  • N-terminal 6xHis tag for facile purification or detection.
  • Low endotoxin (<1 EU/μg) for sensitive biological applications.
  • Verified enzymatic activity (148.432 U/mg) for functional studies.
  • Lyophilized formulation with stabilizers for improved shelf life.
  • Expressed in E. coli to provide consistent recombinant supply.
  • Reconstitution guidance: avoid concentrations below 100 μg/mL in ddH2O.