Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SOD1 Human N-His 50ug
SDP

Catalog No. 5000196777
Encompass_Preferred

Recombinant human superoxide dismutase 1 (SOD1) with an N-terminal 6xHis affinity tag, expressed in Escherichia coli and supplied lyophilized for research use. Suitable for biochemical assays, structural studies, and antibody/antigen applications. High purity and stabilizing excipients facilitate handling and storage.

  • Expressed in Escherichia coli.
  • N-terminal 6xHis affinity tag for purification.
  • Greater than 95% purity by reducing SDS-PAGE.
  • Lyophilized with trehalose and mannitol to enhance stability.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or -20°C long term.
  • UniProt accession P00441 and Gene ID 6647 provided.

Catalog No. 50-001-96777 Supplier Medchemexpress LLC Supplier No. HYP71048A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human superoxide dismutase 1 (SOD1) with an N-terminal 6xHis affinity tag, expressed in Escherichia coli and supplied lyophilized for research use. Suitable for biochemical assays, structural studies, and antibody/antigen applications. High purity and stabilizing excipients facilitate handling and storage.

  • Expressed in Escherichia coli.
  • N-terminal 6xHis affinity tag for purification.
  • Greater than 95% purity by reducing SDS-PAGE.
  • Lyophilized with trehalose and mannitol to enhance stability.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or -20°C long term.
  • UniProt accession P00441 and Gene ID 6647 provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.