Learn More
Medchemexpress LLC SOD1 Human N-His 5ug
SDP
Catalog No. 5000197376
Encompass_Preferred
Recombinant human SOD1 (superoxide dismutase [Cu-Zn] 1) expressed in Escherichia coli with an N-terminal 6xHis tag, supplied in small research quantities (5 μg). Intended for biochemical and immunological research applications including studies of oxidative stress, enzymatic assays, antibody validation, and proteomics.
  • Recombinant human sequence with confirmed accession P00441.
  • N-terminal 6xHis tag facilitates purification and detection.
  • Expressed in Escherichia coli for high-yield production.
  • Available in multiple microgram to milligram sizes for experimental flexibility.
  • Suitable for enzymatic assays, antibody validation, and proteomics workflows.
Catalog No. 50-001-97376 Supplier Medchemexpress LLC Supplier No. HYP71048A5UG
Recombinant human SOD1 (superoxide dismutase [Cu-Zn] 1) expressed in Escherichia coli with an N-terminal 6xHis tag, supplied in small research quantities (5 μg). Intended for biochemical and immunological research applications including studies of oxidative stress, enzymatic assays, antibody validation, and proteomics.
  • Recombinant human sequence with confirmed accession P00441.
  • N-terminal 6xHis tag facilitates purification and detection.
  • Expressed in Escherichia coli for high-yield production.
  • Available in multiple microgram to milligram sizes for experimental flexibility.
  • Suitable for enzymatic assays, antibody validation, and proteomics workflows.