Learn More
ACROBiosystems Recombinant Protein;Human SOD1 / Cu-Zn SOD Protein;E.coli;1MG;SO1,SOD1,ALS,ALS1,HEL-S-44,IPOA,SOD,hSod1,SOD-1,Superoxide dismutase 1
Catalog No. 501658880
Encompass_Preferred
Recombinant Protein;Human SOD1 / Cu-Zn SOD Protein;1MG;Human SOD1, His Tag (SO1-H5148) is expressed from E.coli cells. It contains AA Ala 2 - Gln 154 (Accession # AAH01034).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 20 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8880 Supplier ACROBiosystems Supplier No. SO1H51481MG
Recombinant Protein;Human SOD1 / Cu-Zn SOD Protein;1MG;Human SOD1, His Tag (SO1-H5148) is expressed from E.coli cells. It contains AA Ala 2 - Gln 154 (Accession # AAH01034).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 20 kDa under reducing (R) condition (SDS-PAGE).