Learn More
Sino Biological SOD2 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 12656R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SOD2 (rh SOD2; Catalog#12656-HNAE; P04179-1; Lys 25-Lys 222).
Catalog No. 50-224-7913